2-Nitro-4,5-xylidine
Supplier: Apollo Scientific
|
Danger
|
OR0414-100G
OR0414-25G
OR0414-5G
APOSOR0414-100GEA
171
EUR
APOSOR0414-100G
APOSOR0414-25G
APOSOR0414-5G
2-Nitro-4,5-xylidine
2-Nitro-4,5-xylidine
Formula:
C₈H₁₀N₂O₂ Melting Pt: 139…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007811 CAS Number: 6972-71-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...