1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Supplier: Apollo Scientific
Danger
OR0419-1G OR0419-250MG
APOSOR0419-1GEA 58 EUR
APOSOR0419-1G APOSOR0419-250MG
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Formula: C₆H₇ClN₂O
Boiling Pt: 100 °C/20mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00833258
CAS Number: 55458-67-8
UN: 3261
ADR: 8,II