1,4-Dioxaspiro[4.5]decan-8-one
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal
OR0427-100G OR0427-500G
APOSOR0427-100GEA 69.6 EUR
APOSOR0427-100G APOSOR0427-500G
1,4-Dioxaspiro[4.5]decan-8-one
1,4-Dioxaspiro[4.5]decan-8-one
Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8