3,3-Dimethylbutyric acid
Supplier: Apollo Scientific
Danger
Synonyms: 3,3-Dimethylbutanoic acid, tert-Butylacetic acid
OR0494-100G OR0494-500G OR0494-1KG
APOSOR0494-100GEA 26.1 EUR
APOSOR0494-100G APOSOR0494-500G APOSOR0494-1KG
3,3-Dimethylbutyric acid
3,3-Dimethylbutyric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 185…190 °C
Melting Pt: –11 °C
Density: 0.9124 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002715
CAS Number: 1070-83-3
EINECS: 213-982-2
UN: 3265
ADR: 8,III