2-Formylimidazole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1H-Imidazole-2-carbaldehyde, Imidazole-2-carboxaldehyde
OR0497-100G OR0497-25G
APOSOR0497-100GEA 270 EUR
APOSOR0497-100G APOSOR0497-25G
2-Formylimidazole 95%
2-Formylimidazole
Formula: C₄H₄N₂O
MW: 96.09 g/mol
Boiling Pt: 80 °C/0.1mmHg
Melting Pt: 205…209 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00003544
CAS Number: 10111-08-7