Homopiperazine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1,4-Diazepane, 1,4-Diazacycloheptane, Perhydro-1,4-diazepine
OR0503-100G OR0503-25G
APOSOR0503-100GEA 102 EUR
APOSOR0503-100G APOSOR0503-25G
Homopiperazine 97%
Homopiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 168…170 °C
Melting Pt: 38…40 °C
Density: 0.95 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00006933
CAS Number: 505-66-8
EINECS: 208-016-1
UN: 3259
ADR: 8,III