3-(2-Hydroxyethyl)indole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
OR0519-5G OR0519-25G OR0519-100G
APOSOR0519-5GEA 75.4 EUR
APOSOR0519-5G APOSOR0519-25G APOSOR0519-100G
3-(2-Hydroxyethyl)indole 98%
3-(2-Hydroxyethyl)indole
Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 174 °C/2mmHg
Melting Pt: 57…60 °C
Flash Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00005659
CAS Number: 526-55-6
EINECS: 208-393-2