4'-Hydroxy-3'-methylacetophenone
Supplier: Apollo Scientific
Warning
OR0526-100G OR0526-25G OR0526-5G
APOSOR0526-100GEA 140 EUR
APOSOR0526-100G APOSOR0526-25G APOSOR0526-5G
4'-Hydroxy-3'-methylacetophenone
4'-Hydroxy-3'-methylacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 175 °C/1mmHg
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00002231
CAS Number: 876-02-8
EINECS: 212-880-5