1-Adamantanemethanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol
OR0527-100G OR0527-25G OR0527-500G
APOSOR0527-100GEA 142 EUR
APOSOR0527-100G APOSOR0527-25G APOSOR0527-500G
1-Adamantanemethanol 98%
1-Adamantanemethanol
Formula: C₁₁H₁₈O
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00074751
CAS Number: 770-71-8