5-Mercapto-4-pentyl-4H-1,2,4-triazol-3-ol
Supplier: Apollo Scientific
Warning
Synonyms: 5-Hydroxy-4-pentyl-1,2,4-triazole-3-thiol
OR0536-1G OR0536-5G
APOSOR0536-1GEA 89.9 EUR
APOSOR0536-1G APOSOR0536-5G
5-Mercapto-4-pentyl-4H-1,2,4-triazol-3-ol
5-Mercapto-4-pentyl-4H-1,2,4-triazol-3-ol
Formula: C₇H₁₃N₃OS
MW: 187.27 g/mol
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00207999
CAS Number: 117987-05-0