4-Iodo-2-nitrotoluene 98%
Supplier: Apollo Scientific
Warning
OR0542-100G OR0542-25G OR0542-5G
APOSOR0542-100GEA 103 EUR
APOSOR0542-100G APOSOR0542-25G APOSOR0542-5G
4-Iodo-2-nitrotoluene 98%
4-Iodo-2-nitrotoluene
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00051090
CAS Number: 41252-97-5