2-(4-Isobutylphenyl)propanohydrazide
Supplier: Apollo Scientific
OR0556-1G OR0556-5G OR0556-10G
APOSOR0556-1GEA 132 EUR
APOSOR0556-1G APOSOR0556-5G APOSOR0556-10G
2-(4-Isobutylphenyl)propanohydrazide
2-(4-Isobutylphenyl)propanohydrazide
Formula: C₁₃H₂₀N₂O
MW: 220.32 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00711879
CAS Number: 127222-69-9