4-(Methylsulphonyl)phenol
Supplier: Apollo Scientific
Warning
OR0562-100G OR0562-25G OR0562-5G
APOSOR0562-100GEA 198 EUR
APOSOR0562-100G APOSOR0562-25G APOSOR0562-5G
4-(Methylsulphonyl)phenol
4-(Methylsulphonyl)phenol
Formula: C₇H₈O₃S
MW: 172.20 g/mol
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00047806
CAS Number: 14763-60-1