(Chloromethyl)pentamethylbenzene 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Pentamethylbenzyl chloride, 2,3,4,5,6-Pentamethylbenzyl chloride
OR0567-5G
OR0567-1G
APOSOR0567-5GEA
290
EUR
APOSOR0567-5G
APOSOR0567-1G
(Chloromethyl)pentamethylbenzene 95%
(Chloromethyl)pentamethylbenzene
Formula:
(C₆(CH₃)₅)CH₂Cl Boiling Pt: 140…142 °C/0.8mmHg Melting Pt: 81…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000900 CAS Number: 484-65-1 EINECS: 207-608-7 UN: 3261 ADR: 8,III |