cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione
Supplier: Apollo Scientific
Warning
OR0607-1G OR0607-5G OR0607-250MG
APOSOR0607-1GEA 48 EUR
APOSOR0607-1G APOSOR0607-5G APOSOR0607-250MG
cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione
cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 228…230 °C
Storage Temperature: Ambient
CAS Number: 21170-10-5
EINECS: 244-255-8