2-Methylindan-1-one
Supplier: Apollo Scientific
Synonyms: 2-Methyl-1-indanone
OR0627-25G OR0627-5G
APOSOR0627-25GEA 415 EUR
APOSOR0627-25G APOSOR0627-5G
2-Methylindan-1-one
2-Methylindan-1-one
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 93…95 °C/4mmHg
Density: 1.064 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00192303
CAS Number: 17496-14-9