5-Bromobenzo[b]furan-2-carbaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromobenzofuran-2-carbaldehyde
OR0646-1G
OR0646-250MG
APOSOR0646-1GEA
144
EUR
APOSOR0646-1G
APOSOR0646-250MG
5-Bromobenzo[b]furan-2-carbaldehyde 97%
5-Bromobenzo[b]furan-2-carbaldehyde
Formula:
C₉H₅BrO₂ MW: 225.05 g/mol Melting Pt: 129…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659072 CAS Number: 23145-16-6 |