3-Nitro-4-(1-pyrrolidinyl)benzenecarbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3-Nitro-4-(pyrrolidin-1-yl)benzaldehyde
OR0689-100MG OR0689-500MG
APOSOR0689-100MGEA 21.9 EUR
APOSOR0689-100MG APOSOR0689-500MG
3-Nitro-4-(1-pyrrolidinyl)benzenecarbaldehyde
3-Nitro-4-(1-pyrrolidinyl)benzenecarbaldehyde
Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00455104
CAS Number: 284679-97-6