4-Phenyl-2-thiazoleacetonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Phenylthiazol-2-yl)acetonitrile
OR0711-1G OR0711-250MG OR0711-5G
APOSOR0711-1GEA 126 EUR
APOSOR0711-1G APOSOR0711-250MG APOSOR0711-5G
4-Phenyl-2-thiazoleacetonitrile
4-Phenyl-2-thiazoleacetonitrile
Formula: C₁₁H₈N₂S
MW: 200.259 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00551791
CAS Number: 41381-89-9