4-(Methylthio)phenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Methylmercapto)phenol
OR0778-100G OR0778-50G OR0778-500G
APOSOR0778-100GEA 203 EUR
APOSOR0778-100G APOSOR0778-50G APOSOR0778-500G
4-(Methylthio)phenol 98%
4-(Methylthio)phenol
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 153…156 °C/20mmHg
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002351
CAS Number: 1073-72-9
EINECS: 214-031-4