3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole
Supplier: Apollo Scientific
Warning
OR0783-1G OR0783-5G
APOSOR0783-1GEA 94.4 EUR
APOSOR0783-1G APOSOR0783-5G
3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole
3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole
Formula: C₉H₇BrN₂O
Melting Pt: 98…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00487341
CAS Number: 118183-92-9