1-Carbamoylpiperidine-4-carboxylic acid
Supplier: Apollo Scientific
Warning
OR0808-1G OR0808-250MG OR0808-5G
APOSOR0808-1GEA 114 EUR
APOSOR0808-1G APOSOR0808-250MG APOSOR0808-5G
1-Carbamoylpiperidine-4-carboxylic acid
Formula: C₇H₁₂N₂O₃
Melting Pt: 196 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06655891
CAS Number: 851168-77-9