3-Iodobenzonitrile 98%
Supplier: Apollo Scientific
Warning
OR0825-100G OR0825-25G OR0825-5G
APOSOR0825-100GEA 144 EUR
APOSOR0825-100G APOSOR0825-25G APOSOR0825-5G
3-Iodobenzonitrile 98%
3-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Melting Pt: 39…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079762
CAS Number: 69113-59-3
UN: 3439
ADR: 6.1,III