3-Thiophenecarbonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile
OR0832-25G OR0832-5G
APOSOR0832-25GEA 46.4 EUR
APOSOR0832-25G APOSOR0832-5G
3-Thiophenecarbonitrile 97%
3-Thiophenecarbonitrile
Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 205 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 85.56 °C
Storage Temperature: Ambient
MDL Number: MFCD00151852
CAS Number: 1641-09-4
EINECS: 216-687-7