2-Iodobenzoic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: o-Iodobenzoic acid
OR0837-100G OR0837-25G
APOSOR0837-100GEA 27.7 EUR
APOSOR0837-100G APOSOR0837-25G
2-Iodobenzoic acid 98%
2-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 314
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00002419
CAS Number: 88-67-5
EINECS: 201-850-7
Merck Index: 13,05052