Methyl-3-cyclohexene-1-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-cyclohexenecarboxylate, 3-Cyclohexene-1-carboxylic Acid Methyl Ester, Methyl cyclohex-3-enecarboxylate
OR0842-10G OR0842-1G
APOSOR0842-10GEA 24.8 EUR
APOSOR0842-10G APOSOR0842-1G
Methyl-3-cyclohexene-1-carboxylate 97%
Methyl-3-cyclohexene-1-carboxylate
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 185 °C
Density: 1.02 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00053266
CAS Number: 6493-77-2
EINECS: 229-376-6
UN: 3272
ADR: 3,III