Cyclododecanol 99%
Supplier: Apollo Scientific
Warning
OR0848-100G OR0848-500G
APOSOR0848-100GEA 101 EUR
APOSOR0848-100G APOSOR0848-500G
Cyclododecanol 99%
Cyclododecanol
Formula: C₁₂H₂₄O
Boiling Pt: 273 °C
Melting Pt: 76…79 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00003717
CAS Number: 1724-39-6
EINECS: 217-031-2
UN: 3077
ADR: 9+EHS,III