N-Chlorosuccinimide 99%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Chloropyrrolidine-2,5-dione
OR0856-1KG OR0856-500G
APOSOR0856-1KGEA 72.5 EUR
APOSOR0856-1KG APOSOR0856-500G
N-Chlorosuccinimide 99%
N-Chlorosuccinimide
Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
Melting Pt: 148…150 °C
Density: 1.65 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II