1-(3-Methyl-2-pyridinyl)piperazine
Supplier: Apollo Scientific
|
Warning
|
OR0888-1G
OR0888-250MG
APOSOR0888-1GEA
215
EUR
APOSOR0888-1G
APOSOR0888-250MG
1-(3-Methyl-2-pyridinyl)piperazine
1-(3-Methyl-2-pyridinyl)piperazine
Formula:
C₁₀H₁₅N₃ Boiling Pt: 97 °C/0.5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03412117 CAS Number: 104396-10-3 |