1-(4-Methylpyridin-2-yl)piperazine 97%
Supplier: Apollo Scientific
Danger
OR0889-1G OR0889-250MG OR0889-5G
APOSOR0889-1GEA 134 EUR
APOSOR0889-1G APOSOR0889-250MG APOSOR0889-5G
1-(4-Methylpyridin-2-yl)piperazine 97%
1-(4-Methylpyridin-2-yl)piperazine
Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
Boiling Pt: 161…163 °C/8mmHg
Storage Temperature: Ambient
MDL Number: MFCD03412118
CAS Number: 34803-67-3
UN: 3259
ADR: 8 III