5-Chloro-2-nitrobenzoic acid 98%
Supplier: Apollo Scientific
Warning
OR0907-100G OR0907-500G OR0907-1KG
APOSOR0907-100GEA 34.8 EUR
APOSOR0907-100G APOSOR0907-500G APOSOR0907-1KG
5-Chloro-2-nitrobenzoic acid 98%
5-Chloro-2-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Melting Pt: 136…140 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007290
CAS Number: 2516-95-2
EINECS: 219-738-1