(3-Carboxypropyl)triphenylphosphonium bromide 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Carboxypropyltriphenylphosphonium bromide
OR0912-100G OR0912-25G
APOSOR0912-100GEA 72.5 EUR
APOSOR0912-100G APOSOR0912-25G
(3-Carboxypropyl)triphenylphosphonium bromide 98+%
(3-Carboxypropyl)triphenylphosphonium bromide
Formula: C₂₂H₂₂BrO₂P
Melting Pt: 244…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00274196
CAS Number: 17857-14-6