3,5-Dichlorobenzoic acid
Supplier: Apollo Scientific
Warning
OR0935-100G OR0935-25G
APOSOR0935-100GEA 24.8 EUR
APOSOR0935-100G APOSOR0935-25G
3,5-Dichlorobenzoic acid
3,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00002494
CAS Number: 51-36-5
EINECS: 200-092-4