2-Bromo-5-nitropyridine
Supplier: Apollo Scientific
Danger
OR0936-100G OR0936-25G
APOSOR0936-100GEA 103 EUR
APOSOR0936-100G APOSOR0936-25G
2-Bromo-5-nitropyridine
2-Bromo-5-nitropyridine
Formula: C₅H₃BrN₂O₂
Boiling Pt: 145…147 °C/10mmHg
Melting Pt: 139…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006222
CAS Number: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1,III