Ethyl-4-(1-piperazinyl)benzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4-(piperazin-1-yl)benzoate
OR0944-100G OR0944-25G OR0944-5G
APOSOR0944-100GEA 429 EUR
APOSOR0944-100G APOSOR0944-25G APOSOR0944-5G
Ethyl-4-(1-piperazinyl)benzoate 98%
Ethyl-4-(1-piperazinyl)benzoate
Formula: C₁₃H₁₈N₂O₂
MW: 234.30 g/mol
Melting Pt: 89…90 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04973340
CAS Number: 80518-57-6