3-Chloro-1-propanol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloropropan-1-ol, 1-Chloro-3-hydroxypropane
OR0977-100G OR0977-25G OR0977-500G
APOSOR0977-100GEA 68.3 EUR
APOSOR0977-100G APOSOR0977-25G APOSOR0977-500G
3-Chloro-1-propanol
3-Chloro-1-propanol
Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 160…162 °C
Melting Pt: –20 °C
Density: 1.131 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00002943
CAS Number: 627-30-5
EINECS: 210-992-9
UN: 2849
ADR: 6.1 III