2,4-Dichlorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4-Dichlorobenzene-1-sulphonyl chloride
OR1000-100G OR1000-10G OR1000-25G
APOSOR1000-100GEA 122 EUR
APOSOR1000-100G APOSOR1000-10G APOSOR1000-25G
2,4-Dichlorobenzenesulphonyl chloride 98%
2,4-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Melting Pt: 50…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00052712
CAS Number: 16271-33-3
UN: 3261
ADR: 8,II