1-Methylimidazole (NMI) 99%
Supplier: Apollo Scientific
Danger
Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
OR10004-1L OR10004-2.5L
APOSOR10004-1LEA 72.5 EUR
APOSOR10004-1L APOSOR10004-2.5L
1-Methylimidazole (NMI) 99%
1-Methylimidazole
Formula: C₄H₆N₂
Boiling Pt: 198 °C
Melting Pt: –6 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
EINECS: 210-484-7
UN: 2922
ADR: 8+6.1,II