Isobutyryl chloride 98%
Supplier: Apollo Scientific
Danger
OR10005-100G OR10005-500G OR10005-25G
APOSOR10005-100GEA 43.5 EUR
APOSOR10005-100G APOSOR10005-500G APOSOR10005-25G
Isobutyryl chloride 98%
Isobutyryl chloride
Formula: C₄H₇ClO
MW: 106.55 g/mol
Boiling Pt: 91…93 °C
Melting Pt: –90 °C
Density: 1.017 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3+8,II