4-Pyridone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone
OR1002-25G
OR1002-10G
APOSOR1002-25GEA
24.8
EUR
APOSOR1002-25G
APOSOR1002-10G
4-Pyridone
4-Pyridone
Formula:
C₅H₅NO MW: 95.1 g/mol Boiling Pt: 230…235 °C/12mmHg Melting Pt: 140…147 °C Flash Pt: 221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006419 CAS Number: 626-64-2 EINECS: 210-958-3 |