4-Methylbenzylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Aminomethyl)toluene
OR10024-100G
APOSOR10024-100GEA 121 EUR
APOSOR10024-100G
4-Methylbenzylamine 98%
4-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 195…197 °C
Melting Pt: 41609 °C
Density: 0.955 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008123
CAS Number: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8,III