Guaiacol 98%
Supplier: Apollo Scientific
Warning
APOSOR10036-1KGEA 193 EUR
APOSOR10036-1KG APOSOR10036-5KG APOSOR10036-500G
Guaiacol 98%
Guaiacol
Formula: C₇H₈O₂
MW: 124,14 g/mol
Boiling Pt: 204…206 °C
Melting Pt: 26…31 °C
Density: 1,129 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
EINECS: 201-964-7