5-Nonanone
Supplier: Apollo Scientific
Warning
Synonyms: Nonan-5-one
OR10037-250G OR10037-500G OR10037-100G
APOSOR10037-250GEA 150 EUR
APOSOR10037-250G APOSOR10037-500G APOSOR10037-100G
5-Nonanone
5-Nonanone
Formula: C₉H₁₈O
Boiling Pt: 186…187 °C
Melting Pt: –50 °C
Density: 0.826 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00009484
CAS Number: 502-56-7
EINECS: 207-946-5