2,4-Pentanediol
Supplier: Apollo Scientific
Warning
OR10041-10G OR10041-50G OR10041-500G
APOSOR10041-10GEA 21.9 EUR
APOSOR10041-10G APOSOR10041-50G APOSOR10041-500G
2,4-Pentanediol
2,4-Pentanediol
Formula: C₅H₁₂O₂
Boiling Pt: 200…201 °C
Density: 0.955 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004549
CAS Number: 625-69-4
EINECS: 210-907-5