L(+)-Diisopropyl tartrate
Supplier: Apollo Scientific
Warning
Synonyms: Diisopropyl (2R,3R)-2,3-dihydroxysuccinate, L-(+)-Tartaric acid diisopropyl ester
OR10051-100G OR10051-250G OR10051-500G
APOSOR10051-100GEA 42.2 EUR
APOSOR10051-100G APOSOR10051-250G APOSOR10051-500G
L(+)-Diisopropyl tartrate
L(+)-Diisopropyl tartrate
Asymmetric synthesis catalyst; Chiral intermediate for pharmaceuticals and agrochemicals.
Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064450
CAS Number: 2217-15-4