Tetrabutylammonium iodide
Supplier: Apollo Scientific
Warning
Synonyms: Tetra-n-butylammonium iodide, TBAI
OR10056-100G OR10056-25G
APOSOR10056-100GEA 24.8 EUR
APOSOR10056-100G APOSOR10056-25G
Tetrabutylammonium iodide
Tetrabutylammonium iodide
Formula: C₁₆H₃₆IN
MW: 369.37 g/mol
Melting Pt: 142…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5