Acridine
Supplier: Apollo Scientific
Warning
OR10069-5G OR10069-10G OR10069-1G
APOSOR10069-5GEA 94.4 EUR
APOSOR10069-5G APOSOR10069-10G APOSOR10069-1G
Acridine
Acridine
Formula: C₁₃H₉N
MW: 179.22 g/mol
Boiling Pt: 346 °C
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005025
CAS Number: 260-94-6
EINECS: 205-971-6
UN: 2713
ADR: 6.1 III