Pentylamine
Supplier: Apollo Scientific
Danger
Synonyms: Amylamine, 1-Aminopentane, n-Amylamine, n-Pentylamine, 1-Pentylamine
OR10079-100G OR10079-25G
APOSOR10079-100GEA 154 EUR
APOSOR10079-100G APOSOR10079-25G
Pentylamine
Pentylamine
Formula: H₃C(CH₂)₃CH₂NH₂
MW: 87.16 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –50 °C
Density: 0.752 g/cm³ (25 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008236
CAS Number: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3+8,II