5-Aminopentan-1-ol 50% in aqueous solution
Supplier: Apollo Scientific
Danger
Synonyms: 5-Amino-1-pentanol
OR10081-100G OR10081-25G
APOSOR10081-100GEA 150 EUR
APOSOR10081-100G APOSOR10081-25G
5-Aminopentan-1-ol 50% in aqueous solution
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 122 °C/16mmHg
Melting Pt: 35…37 °C
Density: 0.949 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
UN: 2735
ADR: 8 III