Cyclopropyl methyl ketone
Supplier: Apollo Scientific
Danger
Synonyms: 1-Cyclopropylethanone
OR10100-100G OR10100-25G OR10100-500G
APOSOR10100-100GEA 65.4 EUR
APOSOR10100-100G APOSOR10100-25G APOSOR10100-500G
Cyclopropyl methyl ketone
Cyclopropyl methyl ketone
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 113…114 °C
Melting Pt: < –70 °C
Density: 0.898 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00001297
CAS Number: 765-43-5
EINECS: 212-146-4
UN: 1992
ADR: 3+6.1,II